Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylpiperidine 99%, ACROS Organics™

Click to view available options
Quantity:
10mL
250mL
1L
Packaging:
Glass bottle
626-67-5
C6H13N
99.18
MFCD00006491
PAMIQIKDUOTOBW-UHFFFAOYSA-N
n-methylpiperidine,piperidine, 1-methyl,methylpiperidine,n-methyl piperidine,1-methyl-piperidine,n-methylpiperidin,methylpiperdine,1-methylpiperadine,n-methyl-piperidine,1-methyl piperidine
12291
1-methylpiperidine
CN1CCCCC1
626-67-5
C6H13N
99.18
MFCD00006491
PAMIQIKDUOTOBW-UHFFFAOYSA-N
n-methylpiperidine,piperidine, 1-methyl,methylpiperidine,n-methyl piperidine,1-methyl-piperidine,n-methylpiperidin,methylpiperdine,1-methylpiperadine,n-methyl-piperidine,1-methyl piperidine
12291
1-methylpiperidine
CN1CCCCC1
Viscosity 5 mPa.s (20°C)
Specific Gravity 0.82
Packaging Glass bottle
Quantity 250mL
Boiling Point 105.0°C to 108.0°C
Color Colorless
Melting Point -50.0°C
Infrared Spectrum Authentic
Density 0.8200g/mL
Flash Point 3°C
Refractive Index 1.4368 to 1.4388
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 20,16
Solubility Information Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, acetone, toluene and
Formula Weight 99.18
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material N-Methylpiperidine, 99%
Show More Show Less

GHS P Statement
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger

EINECSNumber : 210-959-9

RTECSNumber : TN1225000

TSCA : TSCA