Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylpyrrole-2-boronic acid pinacol ester, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
850567-47-4
C11H18BNO2
207.08
OEQQEXKRORJZPR-UHFFFAOYSA-N
1-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole,1-methyl-1h-pyrrole-2-boronic acid, pinacol ester,1-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrrole,1-methyl-2-pyrroleboronic acid pinacol ester,n-methylpyrrole-2-boronic acid, pinacol ester,1h-pyrrole, 1-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,pubchem18528,1-methylpyrrole-2-boronic acid pinacol ester,n-methyl-2-pyrroleboronic acid, pinacol ester,1-methyl-2-tetramethyl-1,3,2-dioxaborolan-2-yl pyrrole
44118773
1-methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrole
B1(OC(C(O1)(C)C)(C)C)C2=CC=CN2C
850567-47-4
C11H18BNO2
207.08
OEQQEXKRORJZPR-UHFFFAOYSA-N
1-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole,1-methyl-1h-pyrrole-2-boronic acid, pinacol ester,1-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrrole,1-methyl-2-pyrroleboronic acid pinacol ester,n-methylpyrrole-2-boronic acid, pinacol ester,1h-pyrrole, 1-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,pubchem18528,1-methylpyrrole-2-boronic acid pinacol ester,n-methyl-2-pyrroleboronic acid, pinacol ester,1-methyl-2-tetramethyl-1,3,2-dioxaborolan-2-yl pyrrole
44118773
1-methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrole
B1(OC(C(O1)(C)C)(C)C)C2=CC=CN2C
Melting Point 47°C to 49°C
Color Brown to White
Infrared Spectrum Authentic
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 207.08
Physical Form Powder or Low Melting Solid
Percent Purity 97%
Chemical Name or Material N-Methylpyrrole-2-boronic acid pinacol ester
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning