Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylpyrrole, 99+%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
96-54-8
C5H7N
81.12
MFCD00005345
OXHNLMTVIGZXSG-UHFFFAOYSA-N
1-methyl-1h-pyrrole,n-methylpyrrole,n-methyl pyrrole,1h-pyrrole, 1-methyl,pyrrole, 1-methyl,methylpyrrole,n-methylpyrrol,1-methylpyrrol,unii-ng5gpn98zg,ccris 2934
7304
1-methylpyrrole
CN1C=CC=C1
96-54-8
C5H7N
81.12
MFCD00005345
OXHNLMTVIGZXSG-UHFFFAOYSA-N
1-methyl-1h-pyrrole,n-methylpyrrole,n-methyl pyrrole,1h-pyrrole, 1-methyl,pyrrole, 1-methyl,methylpyrrole,n-methylpyrrol,1-methylpyrrol,unii-ng5gpn98zg,ccris 2934
7304
1-methylpyrrole
CN1C=CC=C1
Quantity 500g
Refractive Index 1.4875
Water 0.5% max. (K.F.)
Density 0.9100g/mL
Melting Point -57°C
Boiling Point 111°C to 114°C
Flash Point 16°C
Specific Gravity 0.91
Viscosity 0.7 mPa.s (20°C)
Infrared Spectrum Authentic
Color Yellow to Brown
Packaging Glass bottle
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Beilstein 20, 163
Solubility Information Solubility in water: 13g/L water (20°C). Other solubilities: soluble in alcohol,miscible with most usual organic solents
Formula Weight 81.12
Physical Form Liquid
Percent Purity ≥99.0%
Chemical Name or Material N-Methylpyrrole, 99+%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-513-7

RTECSNumber : UX9640000

TSCA : TSCA