Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Burdick & Jackson™ N-Methylpyrrolidone, Anhydrous, septum-sealed, for applications requiring low water content, >99.5%, Honeywell Burdick & Jackson

Click to view available options
Quantity:
6 x 100mL
6 x 1L
Packaging:
Septum Sealed Bottle
872-50-4
C5H9NO
99.133
SECXISVLQFMRJM-UHFFFAOYSA-N
1-methyl-2-pyrrolidinone,n-methyl-2-pyrrolidone,n-methylpyrrolidone,1-methyl-2-pyrrolidone,methylpyrrolidone,m-pyrol,n-methyl-2-pyrrolidinone,n-methylpyrrolidinone,1-methylpyrrolidinone,2-pyrrolidinone, 1-methyl
13387
CHEBI:7307
1-methylpyrrolidin-2-one
CN1CCCC1=O
872-50-4
C5H9NO
99.133
SECXISVLQFMRJM-UHFFFAOYSA-N
1-methyl-2-pyrrolidinone,n-methyl-2-pyrrolidone,n-methylpyrrolidone,1-methyl-2-pyrrolidone,methylpyrrolidone,m-pyrol,n-methyl-2-pyrrolidinone,n-methylpyrrolidinone,1-methylpyrrolidinone,2-pyrrolidinone, 1-methyl
13387
CHEBI:7307
1-methylpyrrolidin-2-one
CN1CCCC1=O

Anhydrous, septum-sealed, for applications requiring low water content, >99.5%

  • Water content ranging from <10 ppm to <50 ppm
  • Low residue
  • Convenient and effective packaging ensures product quality
  • Certificate of Analysis printed on the label

 

Quantity 6 x 1L
Packaging Septum Sealed Bottle
CAS Max % 100.0000%
Beilstein 106420
Physical Form anhydrous
Percent Purity >99.5%
Chemical Name or Material N-Methylpyrrolidone, Anhydrous, septum-sealed, for applications requiring low water content, >99.5%

ShelfLife : 365 days from date of manufacture

DOTInformation : Packing Group: 3