Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylquipazine dimaleate, Tocris Bioscience™

Click to view available options
Quantity:
100mg
This item is not returnable. View return policy
171205-17-7
C22H25N3O8
459.46
MFCD00153869
HNSITEGFVDCKMF-UHFFFAOYSA-N
n-methylquipazine dimaleate, unii-3he4v6nq2i, 3he4v6nq2i, n-methylquipazine dimaleate salt, 2-1-4-methyl piperazinyl quinoline dimaleate salt, quinoline, 2-4-methyl-1-piperazinyl-, 2z-2-butenedioate 1:2, quinoline, 2-4-methyl-1-piperazinyl-, z-2-butenedioate 1:2, n-methylquipazine dimaleate salt, solid, n-methylquipazine; bis maleic acid, 2-1-n-methylpiperazinyl quinoline dimaleate
11957670
CHEBI:64162
2-(4-methylpiperazin-1-yl)quinoline; bis(but-2-enedioic acid)
OC(=O)C=CC(O)=O.OC(=O)C=CC(O)=O.CN1CCN(CC1)C1=CC=C2C=CC=CC2=N1
This item is not returnable. View return policy
171205-17-7
C22H25N3O8
459.46
MFCD00153869
HNSITEGFVDCKMF-UHFFFAOYSA-N
n-methylquipazine dimaleate, unii-3he4v6nq2i, 3he4v6nq2i, n-methylquipazine dimaleate salt, 2-1-4-methyl piperazinyl quinoline dimaleate salt, quinoline, 2-4-methyl-1-piperazinyl-, 2z-2-butenedioate 1:2, quinoline, 2-4-methyl-1-piperazinyl-, z-2-butenedioate 1:2, n-methylquipazine dimaleate salt, solid, n-methylquipazine; bis maleic acid, 2-1-n-methylpiperazinyl quinoline dimaleate
11957670
CHEBI:64162
2-(4-methylpiperazin-1-yl)quinoline; bis(but-2-enedioic acid)
OC(=O)C=CC(O)=O.OC(=O)C=CC(O)=O.CN1CCN(CC1)C1=CC=C2C=CC=CC2=N1

5-HT3 agonist

5-HT3 agonist. Has almost the same affinity for 5-HT3 sites as quipazine but unlike the latter, does not bind to 5-HT1B sites.
Quantity 100mg
Formula Weight 459.45
Percent Purity >98%
Chemical Name or Material N-Methylquipazine dimaleate

Recommended Storage : Store at Room Temperature