Alfa Aesar™ N-Methylthiourea, 98+%

Manufacturer: Alfa Aesar™ A1043736

Quantity 500g

 View more versions of this product

Catalog No. AAA1043736

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 598-52-7
    Molecular Formula C2H6N2S
    Formula Weight 90.15
    MDL Number MFCD00004938
    UN Number UN2811
    Beilstein 506162
    Chemical Name or Material N-Methylthiourea

    Specifications

    Formula Weight 90.15
    Health Hazard 3 GHS P Statement
    P280h-P301+P310-P330-P501a
    Wear protective gloves/clothing.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    pH 7
    Quantity 500g
    UN Number UN2811
    Health Hazard 2 GHS H Statement
    H300-H361
    Fatal if swallowed.
    Suspected of damaging fertility or the unborn child.
    RTECS Number YT9000000
    MDL Number MFCD00004938
    CAS 598-52-7
    Beilstein 506162
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Melting Point 119° to 124°C
    Molecular Formula C2H6N2S
    Chemical Name or Material N-Methylthiourea
    Assay ≥98%

    GHS H Statement
    H300-H361
    Fatal if swallowed.
    Suspected of damaging fertility or the unborn child.

    GHS P Statement
    P280h-P301+P310-P330-P501a
    Wear protective gloves/clothing.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit