Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nω-Methyltryptamine 99%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
61-49-4
C11H14N2
174.247
MFCD00005657
NCIKQJBVUNUXLW-UHFFFAOYSA-N
n-methyltryptamine,3-2-methylaminoethyl indole,2-1h-indol-3-yl-n-methylethanamine,dipterine,methyltryptamine,n-monomethyltryptamine,1h-indole-3-ethanamine, n-methyl,n-omega-methyltryptamine,3-2-methylamino ethyl indole,unii-6frl4l3z7v
6088
CHEBI:28136
2-(1H-indol-3-yl)-N-methylethanamine
CNCCC1=CNC2=CC=CC=C21
61-49-4
C11H14N2
174.247
MFCD00005657
NCIKQJBVUNUXLW-UHFFFAOYSA-N
n-methyltryptamine,3-2-methylaminoethyl indole,2-1h-indol-3-yl-n-methylethanamine,dipterine,methyltryptamine,n-monomethyltryptamine,1h-indole-3-ethanamine, n-methyl,n-omega-methyltryptamine,3-2-methylamino ethyl indole,unii-6frl4l3z7v
6088
CHEBI:28136
2-(1H-indol-3-yl)-N-methylethanamine
CNCCC1=CNC2=CC=CC=C21
Melting Point 87.0°C to 91.0°C
Water 1% max. (K.F.)
Color White
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 174.24
Physical Form Shiny Crystalline Powder or Flakes
Percent Purity 99%
Chemical Name or Material N-ω-Methyltryptamine, 99%
Show More Show Less

EINECSNumber : 200-507-9

RTECSNumber : NM0450000