Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-MPPP Hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
207452-97-9
C21H27ClN2O
358.91
BJJKMJWQLJIRGF-UHFFFAOYSA-N
n-mppp hydrochloride,n-methyl-n-1s-1-phenyl-2-1-pyrrolidinyl ethyl phenylacetamide hydrochloride
53412424
N-methyl-2-phenyl-N-(1-phenyl-2-pyrrolidin-1-ylethyl)acetamide;hydrochloride
CN(C(CN1CCCC1)C2=CC=CC=C2)C(=O)CC3=CC=CC=C3.Cl
207452-97-9
C21H27ClN2O
358.91
BJJKMJWQLJIRGF-UHFFFAOYSA-N
n-mppp hydrochloride,n-methyl-n-1s-1-phenyl-2-1-pyrrolidinyl ethyl phenylacetamide hydrochloride
53412424
N-methyl-2-phenyl-N-(1-phenyl-2-pyrrolidin-1-ylethyl)acetamide;hydrochloride
CN(C(CN1CCCC1)C2=CC=CC=C2)C(=O)CC3=CC=CC=C3.Cl

Selective κ agonist

High affinity κ agonist with no measured binding at μ or δ sites.
Quantity 50mg
Formula Weight 358.91
Percent Purity >98%
Chemical Name or Material N-MPPP Hydrochloride

Recommended Storage : Store at Room Temperature