Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-MPPP Hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

207452-97-9
C21H27ClN2O
358.91
BJJKMJWQLJIRGF-UHFFFAOYSA-N
N-Methyl-N-[(1S)-1-phenyl-2-(1-pyrrolidinyl)ethyl]phenylacetamide hydrochloride
53412424

Selective κ agonist

High affinity κ agonist with no measured binding at μ or δ sites.
Quantity 10mg
SMILES CN(C(CN1CCCC1)C2=CC=CC=C2)C(=O)CC3=CC=CC=C3.Cl
IUPAC Name N-methyl-2-phenyl-N-(1-phenyl-2-pyrrolidin-1-ylethyl)acetamide;hydrochloride
Formula Weight 358.91
Chemical Name or Material N-MPPP Hydrochloride
Grade >98%

Recommended Storage : Store at Room Temperature