Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N myc interactor Mouse anti-Human, Clone: PCRP-NMI-1C1, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen N myc interactor
Clone PCRP-NMI-1C1
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, Protein Array
Applications Flow Cytometry, Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122453
View Documents
Novus Biologicals
NBP30749010UG
100 μg
Each for $352.89
Add to cart
 
NB122454
View Documents
Novus Biologicals
NBP30749020UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

N myc interactor Monoclonal antibody specifically detects N myc interactor in Human samples. It is validated for Flow Cytometry, Protein Array
Specifications

Specifications

N myc interactor
0.2 mg/ml
Flow Cytometry, Peptide Array
Purified
RUO
Human
Nmi, N-myc (and STAT) interactor, N-myc and STAT interactor, N-myc interactor, N-myc-interactor
Recombinant full-length human N myc interactor protein
Monoclonal
Protein A or G purified
PCRP-NMI-1C1
Flow Cytometry 1-2 ug/million cells, Protein Array
Unconjugated
Mouse
Cancer, Signal Transduction, Tumor Suppressors
10 mM PBS with 0.05% BSA
9111
IgG2b
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit