Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Myristoyl-D-erythro-sphingosine, 99+%, synthetical, ACROS Organics™

Click to view available options
Quantity:
50mg
Packaging:
Glass bottle
34227-72-0
C32H63NO3
509.86
MFCD01320386
ZKRPGPZHULJLKJ-JHRQRACZSA-N
c14 ceramide,c14 cer,n-myristoylsphingosine,n-myristoyl-ceramide,n-tetradecanoyl-sphing-4-enine,n-tetradecanoyl-ceramide,n-myristoyl-d-sphingosine,n-tetradecanoyl sphing-4-enine,n-myristoyl ceramide,n-tetradecanoylsphingosine
5282310
CHEBI:72957
N-[(E,2S,3R)-1,3-dihydroxyoctadec-4-en-2-yl]tetradecanamide
CCCCCCCCCCCCCC=CC(C(CO)NC(=O)CCCCCCCCCCCCC)O
34227-72-0
C32H63NO3
509.86
MFCD01320386
ZKRPGPZHULJLKJ-JHRQRACZSA-N
c14 ceramide,c14 cer,n-myristoylsphingosine,n-myristoyl-ceramide,n-tetradecanoyl-sphing-4-enine,n-tetradecanoyl-ceramide,n-myristoyl-d-sphingosine,n-tetradecanoyl sphing-4-enine,n-myristoyl ceramide,n-tetradecanoylsphingosine
5282310
CHEBI:72957
N-[(E,2S,3R)-1,3-dihydroxyoctadec-4-en-2-yl]tetradecanamide
CCCCCCCCCCCCCC=CC(C(CO)NC(=O)CCCCCCCCCCCCC)O
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Quantity 50mg
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Formula Weight 509.86
Physical Form Powder
Percent Purity ≥99.0%
Chemical Name or Material N-Myristoyl-D-erythro-sphingosine, 99+%
Show More Show Less