Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,2,2-Tetramethyl-1,3-propanediamine, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
53369-71-4
C7H18N2
130.23
ULDIVZQLPBUHAG-UHFFFAOYSA-N
n,n,2,2-tetramethyl-1,3-propanediamine,1,3-propanediamine, n,n,2,2-tetramethyl,n,n,2,2-tetramethylpropane-1,3-diamine,n,n-dimethylneopentanediamine,3-amino-2,2-dimethylpropyl dimethylamine,n1,n1,2,2-tetramethylpropane-1,3-diamine,1,3-propanediamine, n1,n1,2,2-tetramethyl,acmc-209l6k,ksc597o9f,uldivzqlpbuhag-uhfffaoysa
104477
N',N',2,2-tetramethylpropane-1,3-diamine
CC(C)(CN)CN(C)C
53369-71-4
C7H18N2
130.23
ULDIVZQLPBUHAG-UHFFFAOYSA-N
n,n,2,2-tetramethyl-1,3-propanediamine,1,3-propanediamine, n,n,2,2-tetramethyl,n,n,2,2-tetramethylpropane-1,3-diamine,n,n-dimethylneopentanediamine,3-amino-2,2-dimethylpropyl dimethylamine,n1,n1,2,2-tetramethylpropane-1,3-diamine,1,3-propanediamine, n1,n1,2,2-tetramethyl,acmc-209l6k,ksc597o9f,uldivzqlpbuhag-uhfffaoysa
104477
N',N',2,2-tetramethylpropane-1,3-diamine
CC(C)(CN)CN(C)C
Boiling Point 155.0°C to 158.0°C
Quantity 100g
Refractive Index 1.4355
Specific Gravity 0.82
Flash Point 35°C
Color Colorless to Yellow
Density 0.8200g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula (CH3)2NCH2C(CH3)2CH2NH2
Beilstein 04,III,596
Formula Weight 130.23
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material N,N,2,2-Tetramethyl-1,3-propanediamine
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER

GHS Signal Word: Danger

EINECSNumber : 258-501-7