Alfa Aesar™ N,N,2,4,6-Pentamethylaniline, 98%

Manufacturer: Alfa Aesar™ A1056106

Quantity 5g

 View more versions of this product

Catalog No. AAA1056106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13021-15-3
    Molecular Formula C11H17N
    Formula Weight 163.27
    MDL Number MFCD00010596
    Synonym 2-Dimethylaminomesitylene
    Chemical Name or Material N,N,2,4,6-Pentamethylaniline

    Specifications

    Formula Weight 163.27
    Health Hazard 3 GHS P Statement
    P210-P261-P301+P310-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 78°C (174°F)
    Molecular Formula C11H17N
    Boiling Point 213° to 215°C
    Quantity 5g
    MDL Number MFCD00010596
    CAS 13021-15-3
    Synonym 2-Dimethylaminomesitylene
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H227-H315-H319-H335
    Toxic if swallowed.
    Combustible liquid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material N,N,2,4,6-Pentamethylaniline
    Assay 98%

    GHS H Statement
    H301-H227-H315-H319-H335
    Toxic if swallowed.
    Combustible liquid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P301+P310-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit