Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-bis(2-Hydroxyethyl)-2-aminoethanesulfonic acid, free acid, MP Biomedicals™

Catalog Number p-4403862
Click to view available options
Quantity:
100 g
250 g
1 kg
10191-18-1
C6H15NO5S
213.25
MFCD00007533
AJTVSSFTXWNIRG-UHFFFAOYSA-N
N, N-bis(2-Hydroxyethyl)-2-aminoethanesulfonic acid, N, N-Bis(2-hydroxyethyl)taurine
73243
CHEBI:39045
2-[bis(2-hydroxyethyl)amino]ethane-1-sulfonic acid
OCCN(CCO)CCS(O)(=O)=O
10191-18-1
C6H15NO5S
213.25
MFCD00007533
AJTVSSFTXWNIRG-UHFFFAOYSA-N
N, N-bis(2-Hydroxyethyl)-2-aminoethanesulfonic acid, N, N-Bis(2-hydroxyethyl)taurine
73243
CHEBI:39045
2-[bis(2-hydroxyethyl)amino]ethane-1-sulfonic acid
OCCN(CCO)CCS(O)(=O)=O
Color White
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 0.9961
Formula Weight 239.23
Chemical Name or Material N, N-bis(2-Hydroxyethyl)-2-aminoethanesulfonic acid, free acid
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit