Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Bis(2-hydroxyethyl)ethylenediamine, 95%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
4439-20-7
C6H16N2O2
148.2
MFCD00002842
GFIWSSUBVYLTRF-UHFFFAOYSA-N
n,n'-bis 2-hydroxyethyl ethylenediamine,n,n'-diethanolethylenediamine,ethanol, 2,2'-1,2-ethanediyldiimino bis,1,2-bis 2-hydroxyethyl amino ethane,3,6-diazaoctane-1,8-diol,2,2'-ethane-1,2-diylbis azanediyl diethanol,n,n'-bis 2-hydroxyethyl ethylene diamine,n,n'-bis 2-hydroxyethyl-ethylenediamine,2-2-2-hydroxyethylamino ethylamino ethanol
78179
2-[2-(2-hydroxyethylamino)ethylamino]ethanol
C(CNCCO)NCCO
4439-20-7
C6H16N2O2
148.2
MFCD00002842
GFIWSSUBVYLTRF-UHFFFAOYSA-N
n,n'-bis 2-hydroxyethyl ethylenediamine,n,n'-diethanolethylenediamine,ethanol, 2,2'-1,2-ethanediyldiimino bis,1,2-bis 2-hydroxyethyl amino ethane,3,6-diazaoctane-1,8-diol,2,2'-ethane-1,2-diylbis azanediyl diethanol,n,n'-bis 2-hydroxyethyl ethylene diamine,n,n'-bis 2-hydroxyethyl-ethylenediamine,2-2-2-hydroxyethylamino ethylamino ethanol
78179
2-[2-(2-hydroxyethylamino)ethylamino]ethanol
C(CNCCO)NCCO
Melting Point 92°C to 99°C
Infrared Spectrum Authentic
Quantity 50g
Packaging Glass bottle
Color White to Yellow
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula (HOCH2CH2NHCH2-)2
Formula Weight 148.2
Physical Form Powder and/or Chunks
Percent Purity 95%
Chemical Name or Material N, N'-Bis(2-hydroxyethyl)ethylenediamine
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 224-656-4