N,N'-Bis(acryloyl)cystamine, 98%, Alfa Aesar™

Used as a reversible cross-linker for polyacrylamide gels

Manufacturer: Alfa Aesar 4413214

Packaging Packaged under Argon
Quantity 25g

 View more versions of this product

Catalog No. AA4413214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 60984-57-8
    Molecular Formula C10H16N2O2S2
    Linear Formula (CH2=CHC(O)NHCH2CH2S)2
    Formula Weight 260.37
    MDL Number MFCD00036225
    Synonym BAC
    Chemical Name or Material N,N'-Bis(acryloyl)cystamine

    Specifications

    Melting Point 123° to 124°C
    Formula Weight 260.37
    Quantity 25g
    Molecular Formula C10H16N2O2S2
    Health Hazard 1 Warning
    Storage Note 1 −30° to −10°C
    Linear Formula (CH2=CHC(O)NHCH2CH2S)2
    MDL Number MFCD00036225
    CAS 60984-57-8
    Synonym BAC
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a;
    Avoid breathing dust/fume/gas/mist/vapours/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Packaging Packaged under Argon
    Color White
    Physical Form Powder
    Health Hazard 2 GHS H Statement
    H315-H319-H335;
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material N,N'-Bis(acryloyl)cystamine
    Assay 98%

    GHS H Statement
    H315-H319-H335;
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a;
    Avoid breathing dust/fume/gas/mist/vapours/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    −30° to −10°C

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit