Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N,N-Bis(Carboxymethyl)-Dl-Alanine Trisodium Salt, ≥90.0%, Honeywell™ Fluka™
Click to view available options
Quantity:
100g
Packaging:
Poly bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 100g |
Packaging | Poly bottle |
UN Number | NONH for all modes of transport |
Formula Weight | 271.11g/mol |
Percent Purity | ≥90.0% |
Chemical Name or Material | N,N-Bis(Carboxymethyl)-Dl-Alanine Trisodium Salt |