Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N,N'-Di(1-naphthyl)benzidine, 98%

Click to view available options
Quantity:
250mg
1g
5g
152670-41-2
C32H24N2
436.558
MFCD09261380
SZDXPEWZZGNIBB-UHFFFAOYSA-N
n,n'-di 1-naphthyl-4,4'-benzidine,n4,n4'-di naphthalen-1-yl-1,1'-biphenyl-4,4'-diamine,n4,n4'-di-naphthalen-1-yl-biphenyl-4,4'-diamine,n4,n4'-di naphthalen-1-yl biphenyl-4,4'-diamine,n,n'-di 1-naphthyl benzidine,1,1'-biphenyl-4,4'-diamine,n4,n4'-di-1-naphthalenyl,n, n'-dinaphthol-benzidine,n4,n4'-bis naphthalen-1-yl-1,1'-biphenyl-4,4'-diamine,pubchem12682,acmc-1bwou
18958605
N-[4-[4-(naphthalen-1-ylamino)phenyl]phenyl]naphthalen-1-amine
C1=CC=C2C(=C1)C=CC=C2NC3=CC=C(C=C3)C4=CC=C(C=C4)NC5=CC=CC6=CC=CC=C65
152670-41-2
C32H24N2
436.558
MFCD09261380
SZDXPEWZZGNIBB-UHFFFAOYSA-N
n,n'-di 1-naphthyl-4,4'-benzidine,n4,n4'-di naphthalen-1-yl-1,1'-biphenyl-4,4'-diamine,n4,n4'-di-naphthalen-1-yl-biphenyl-4,4'-diamine,n4,n4'-di naphthalen-1-yl biphenyl-4,4'-diamine,n,n'-di 1-naphthyl benzidine,1,1'-biphenyl-4,4'-diamine,n4,n4'-di-1-naphthalenyl,n, n'-dinaphthol-benzidine,n4,n4'-bis naphthalen-1-yl-1,1'-biphenyl-4,4'-diamine,pubchem12682,acmc-1bwou
18958605
N-[4-[4-(naphthalen-1-ylamino)phenyl]phenyl]naphthalen-1-amine
C1=CC=C2C(=C1)C=CC=C2NC3=CC=C(C=C3)C4=CC=C(C=C4)NC5=CC=CC6=CC=CC=C65
Quantity 250mg
Beilstein 3484371
Formula Weight 436.56
Physical Form Solid
Percent Purity 98%
Chemical Name or Material N,N'-Di(1-naphthyl)benzidine