Alfa Aesar™ N,N'-Di(o-tolyl)ethylenediamine, 98%

Manufacturer: Alfa Aesar™ B2497114

Quantity 25g

 View more versions of this product

Catalog No. AAB2497114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 94-92-8
    Molecular Formula C16H20N2
    Formula Weight 240.35
    MDL Number MFCD00048073
    Chemical Name or Material N,N'-Di(o-tolyl)ethylenediamine

    Specifications

    Formula Weight 240.35
    Melting Point 70° to 73°C
    Quantity 25g
    Molecular Formula C16H20N2
    MDL Number MFCD00048073
    CAS 94-92-8
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Chemical Name or Material N,N'-Di(o-tolyl)ethylenediamine
    Assay 98%

    GHS H Statement
    H301-H311-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit