Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N,N'-Di(o-tolyl)ethylenediamine, 98%

Click to view available options
Quantity:
5g
25g
100g
94-92-8
C16H20N2
240.35
MFCD00048073
ZQMPWXFHAUDENN-UHFFFAOYSA-N
n,n'-di-o-tolylethylenediamine,n,n'-di o-tolyl ethylenediamine,n,n'-ethylenedi-o-toluidine,1,2-ethanediamine, n,n'-bis 2-methylphenyl,1,2-ethanediamine, n1,n2-bis 2-methylphenyl,n,n'-bis 2-methylphenyl ethane-1,2-diamine,2-methylphenyl 2-2-methylphenyl amino ethyl amine,n1,n2-bis 2-methylphenyl ethane-1,2-diamine,cambridge id 5185196,n,n'-di-o-tolylethylendiamin
66759
N,N'-bis(2-methylphenyl)ethane-1,2-diamine
CC1=CC=CC=C1NCCNC2=CC=CC=C2C
94-92-8
C16H20N2
240.35
MFCD00048073
ZQMPWXFHAUDENN-UHFFFAOYSA-N
n,n'-di-o-tolylethylenediamine,n,n'-di o-tolyl ethylenediamine,n,n'-ethylenedi-o-toluidine,1,2-ethanediamine, n,n'-bis 2-methylphenyl,1,2-ethanediamine, n1,n2-bis 2-methylphenyl,n,n'-bis 2-methylphenyl ethane-1,2-diamine,2-methylphenyl 2-2-methylphenyl amino ethyl amine,n1,n2-bis 2-methylphenyl ethane-1,2-diamine,cambridge id 5185196,n,n'-di-o-tolylethylendiamin
66759
N,N'-bis(2-methylphenyl)ethane-1,2-diamine
CC1=CC=CC=C1NCCNC2=CC=CC=C2C
Quantity 100g
Melting Point 70°C to 73°C
Formula Weight 240.35
Percent Purity 98%
Chemical Name or Material N,N'-Di(o-tolyl)ethylenediamine

GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA