Alfa Aesar™ N,N'-Diacetyl-1,6-diaminohexane, 98+%

Manufacturer: Alfa Aesar™ B2277109

Quantity 10g

 View more versions of this product

Catalog No. AAB2277109

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3073-59-4
    Molecular Formula CH3CONH(CH2)6NHCOCH3
    Formula Weight 200.28
    MDL Number MFCD00008684
    Beilstein 1775764
    Synonym N,N'-Diacetyl-1,6-hexanediamine; N,N'-Hexamethylenebis(acetamide)
    Chemical Name or Material N,N'-Diacetyl-1,6-diaminohexane

    Specifications

    Formula Weight 200.28
    Quantity 10g
    MDL Number MFCD00008684
    Melting Point 126° to 129°C
    Molecular Formula CH3CONH(CH2)6NHCOCH3
    CAS 3073-59-4
    Beilstein 1775764
    Synonym N,N'-Diacetyl-1,6-hexanediamine; N,N'-Hexamethylenebis(acetamide)
    RTECS Number AC2976200
    Chemical Name or Material N,N'-Diacetyl-1,6-diaminohexane
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit