Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Diacetylhydrazine, 98%, Acros Organics

Click to view available options
Quantity:
25g
3148-73-0
C4H8N2O2
116.12
MFCD00008673
ZLHNYIHIHQEHJQ-UHFFFAOYSA-N
n,n'-diacetylhydrazine,1,2-diacetylhydrazine,acetic acid, 2-acetylhydrazide,diacetylahydrazine,diacetylhydrazine,sym-diacetylhydrazine,unii-ymr3zf0q4m,hydrazine, 1,2-diacetyl,1.2-diacetylhydrazine,ymr3zf0q4m
72884
CHEBI:80616
N'-acetylacetohydrazide
CC(=O)NNC(=O)C
3148-73-0
C4H8N2O2
116.12
MFCD00008673
ZLHNYIHIHQEHJQ-UHFFFAOYSA-N
n,n'-diacetylhydrazine,1,2-diacetylhydrazine,acetic acid, 2-acetylhydrazide,diacetylahydrazine,diacetylhydrazine,sym-diacetylhydrazine,unii-ymr3zf0q4m,hydrazine, 1,2-diacetyl,1.2-diacetylhydrazine,ymr3zf0q4m
72884
CHEBI:80616
N'-acetylacetohydrazide
CC(=O)NNC(=O)C
Quantity 25g
Melting Point 137°C to 141°C
Infrared Spectrum Authentic
Color White
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3CONHNHCOCH3
Formula Weight 116.12
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material N, N'-Diacetylhydrazine, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.