Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N,N'-Diallyl-L-tartardiamide, Electrophoresis Reagent, 99%

Click to view available options
Quantity:
50g
250g
58477-85-3
C10H16N2O4
228.248
MFCD00008640
ZRKLEAHGBNDKHM-YUMQZZPRSA-N
n,n'-diallyltartardiamide,2r,3s-2,3-dihydroxy-n,n'-bis prop-2-enyl butanediamide,2r,3s-2,3-dihydroxy-n,n'-bis prop-2-en-1-yl succinamide
6994946
(2S,3S)-2,3-dihydroxy-N,N'-bis(prop-2-enyl)butanediamide
C=CCNC(=O)C(C(C(=O)NCC=C)O)O
58477-85-3
C10H16N2O4
228.248
MFCD00008640
ZRKLEAHGBNDKHM-YUMQZZPRSA-N
n,n'-diallyltartardiamide,2r,3s-2,3-dihydroxy-n,n'-bis prop-2-enyl butanediamide,2r,3s-2,3-dihydroxy-n,n'-bis prop-2-en-1-yl succinamide
6994946
(2S,3S)-2,3-dihydroxy-N,N'-bis(prop-2-enyl)butanediamide
C=CCNC(=O)C(C(C(=O)NCC=C)O)O
Color White
Melting Point 184°C to 186°C
Quantity 50g
Beilstein 1712934
Formula Weight 228.25
Physical Form Flakes
Percent Purity 99%
Chemical Name or Material N,N'-Diallyl-L-tartardiamide, Electrophoresis Reagent