Alfa Aesar™ N,N'-Dibenzoylhydrazine, 98+%

Manufacturer: Alfa Aesar™ B2508106

Quantity 5g

 View more versions of this product

Catalog No. AAB2508106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 787-84-8
    Molecular Formula (C6H5)CONHNHCO(C6H5)
    Formula Weight 240.26
    MDL Number MFCD00003068
    Chemical Name or Material N,N'-Dibenzoylhydrazine

    Specifications

    Formula Weight 240.26
    TSCA TSCA
    Melting Point 238° to 240°C
    Quantity 5g
    Molecular Formula (C6H5)CONHNHCO(C6H5)
    MDL Number MFCD00003068
    CAS 787-84-8
    Chemical Name or Material N,N'-Dibenzoylhydrazine
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit