Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (+)-N,N'-Dibenzyl-L-tartaric diamide

Click to view available options
Quantity:
1g
5g
25g
88393-56-0
C18H20N2O4
328.368
MFCD00064478
BBYSAHVLSFBCMN-HZPDHXFCSA-N
2r,3r-n1,n4-dibenzyl-2,3-dihydroxysuccinamide,2r,3r-n,n'-dibenzyl-2,3-dihydroxysuccinamide,dibenzyl tartramide,n,n'-dibenzyl-l-tartramide,+-n,n-dibenzyl-l-tartaric diamide,+-n,n'-dibenzyl-l-tartaric diamide,2r,3r-n,n'-dibenzyl-2,3-dihydroxybutanediamide,n,n inverted exclamation marka-dibenzyl-l-tartramide,+-n,n inverted exclamation marka-dibenzyl-l-tartaric diamide
7271811
(2R,3R)-N,N'-dibenzyl-2,3-dihydroxybutanediamide
C1=CC=C(C=C1)CNC(=O)C(C(C(=O)NCC2=CC=CC=C2)O)O
88393-56-0
C18H20N2O4
328.368
MFCD00064478
BBYSAHVLSFBCMN-HZPDHXFCSA-N
2r,3r-n1,n4-dibenzyl-2,3-dihydroxysuccinamide,2r,3r-n,n'-dibenzyl-2,3-dihydroxysuccinamide,dibenzyl tartramide,n,n'-dibenzyl-l-tartramide,+-n,n-dibenzyl-l-tartaric diamide,+-n,n'-dibenzyl-l-tartaric diamide,2r,3r-n,n'-dibenzyl-2,3-dihydroxybutanediamide,n,n inverted exclamation marka-dibenzyl-l-tartramide,+-n,n inverted exclamation marka-dibenzyl-l-tartaric diamide
7271811
(2R,3R)-N,N'-dibenzyl-2,3-dihydroxybutanediamide
C1=CC=C(C=C1)CNC(=O)C(C(C(=O)NCC2=CC=CC=C2)O)O
Color White
Melting Point 198°C to 200°C
Quantity 1g
Beilstein 4923000
Formula Weight 328.37
Physical Form Powder
Chemical Name or Material (+)-N,N'-Dibenzyl-L-tartaric diamide