Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Dicyclohexylcarbodiimide, 0.1M sol.in DCM, peptide synthesis grade, ACROS Organics™

Click to view available options
Quantity:
100mL
Packaging:
Glass bottle
538-75-0
C13H22N2
206.333
MFCD00011659
QOSSAOTZNIDXMA-UHFFFAOYSA-N
dicyclohexylcarbodiimide,n,n'-dicyclohexylcarbodiimide,dccd,1,3-dicyclohexylcarbodiimide,dcci,carbodicyclohexylimide,bis cyclohexyl carbodiimide,dcc,n,n-dicyclohexylcarbodiimide,carbodiimide, dicyclohexyl
10868
CHEBI:53090
N,N'-dicyclohexylmethanediimine
C1CCC(CC1)N=C=NC2CCCCC2
538-75-0
C13H22N2
206.333
MFCD00011659
QOSSAOTZNIDXMA-UHFFFAOYSA-N
dicyclohexylcarbodiimide,n,n'-dicyclohexylcarbodiimide,dccd,1,3-dicyclohexylcarbodiimide,dcci,carbodicyclohexylimide,bis cyclohexyl carbodiimide,dcc,n,n-dicyclohexylcarbodiimide,carbodiimide, dicyclohexyl
10868
CHEBI:53090
N,N'-dicyclohexylmethanediimine
C1CCC(CC1)N=C=NC2CCCCC2
Coupling reagent
Concentration 0.09 to 0.11M
Fieser 01,231; 02,126; 03,91; 04,141; 05,206; 06,174; 07,100; 08,162; 09,156; 10,142; 11,173; 13,333; 14,131; 15,131; 16,128
Density 1.3250g/mL
Merck Index 15, 3107
Formula Weight 206.33
Physical Form Solution
Packaging Glass bottle
Color Undesignated
Quantity 100mL
Chemical Name or Material N, N'-Dicyclohexylcarbodiimide

GHS H Statement:
May cause an allergic skin reaction.
Suspected of causing cancer.
Toxic if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger