Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethyl-1,1,2,3,3,3-hexafluoropropylamine, 90%, Acros Organics

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
309-88-6
C7H11F6N
223.16
MFCD00054683
BNTFCVMJHBNJAR-UHFFFAOYSA-N
n,n-diethyl-1,1,2,3,3,3-hexafluoropropylamine,n,n-diethyl 1,1,2,3,3,3-hexafluoropropylamine,diethyl 1,1,2,3,3,3-hexafluoropropyl amine,1,1,2,3,3,3-hexafluoro-1-diethylaminopropane,n,n-diethyl-1,1,2,3,3,3-hexafluoro-1-propanamine,1-propanamine, n,n-diethyl-1,1,2,3,3,3-hexafluoro,n,n-diethyl-1,1,2,3,3,3-hexafluoropropyl amine,ishikawa s reagent,acmc-20ak9c,ishikawa's reagant
136149
N,N-diethyl-1,1,2,3,3,3-hexafluoropropan-1-amine
CCN(CC)C(C(C(F)(F)F)F)(F)F
309-88-6
C7H11F6N
223.16
MFCD00054683
BNTFCVMJHBNJAR-UHFFFAOYSA-N
n,n-diethyl-1,1,2,3,3,3-hexafluoropropylamine,n,n-diethyl 1,1,2,3,3,3-hexafluoropropylamine,diethyl 1,1,2,3,3,3-hexafluoropropyl amine,1,1,2,3,3,3-hexafluoro-1-diethylaminopropane,n,n-diethyl-1,1,2,3,3,3-hexafluoro-1-propanamine,1-propanamine, n,n-diethyl-1,1,2,3,3,3-hexafluoro,n,n-diethyl-1,1,2,3,3,3-hexafluoropropyl amine,ishikawa s reagent,acmc-20ak9c,ishikawa's reagant
136149
N,N-diethyl-1,1,2,3,3,3-hexafluoropropan-1-amine
CCN(CC)C(C(C(F)(F)F)F)(F)F
C-X bond formation reagent
Flash Point 40°C
Specific Gravity 1.23
Density 1.2300g/mL
Quantity 100g
Color Colorless to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90% (Sum of two isomers) Approximate composition (GC)
Linear Formula (CH3CH2)2NCH2F2CHFCF3
Merck Index 15, 5153
Formula Weight 223.16
Physical Form Liquid
Percent Purity 90%
Chemical Name or Material N,N-Diethyl-1,1,2,3,3,3-hexafluoropropylamine
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger