Learn More
N,N-Diethylaniline, 99%, Thermo Scientific Chemicals
Description
N,N-Diethylaniline is used as an intermediate in pharmaceuticals, dyes and other chemicals. It acts as a reaction catalyst. Its complex, diethylaniline borane is used as a reducing agent in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N,N-Diethylaniline is used as an intermediate in pharmaceuticals, dyes and other chemicals. It acts as a reaction catalyst. Its complex, diethylaniline borane is used as a reducing agent in organic synthesis.
Solubility
Miscible with water and acetone. Slightly miscible with chloroform, alcohol and ether.
Notes
Incompatible with strong oxidizing agents and strong acids. .
Specifications
Melting Point | -38°C |
Density | 0.936 |
Boiling Point | 215°C to 216°C |
Flash Point | 85°C (185°F) |
Refractive Index | 1.542 |
Quantity | 10,000 mL |
UN Number | UN2432 |
Beilstein | 742483 |
Merck Index | 14,3114 |
Solubility Information | Miscible with water and acetone. Slightly miscible with chloroform,alcohol and ether. |
Show More |
Safety and Handling
Hazard Category | H227-H301+H311+H331-H373 |
Hazard Statement | GHS H Statement H301-H311-H331-H373-H227 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Combustible liquid. |
Precautionary Statement | P210-P235-P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: N,N DIETHYLANILINE |
EINECSNumber | 202-088-8 |
RTECSNumber | BX3400000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.