Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethylaniline, 99%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
Packaging:
Glass bottle
91-66-7
C10H15N
149.24
MFCD00009042
GGSUCNLOZRCGPQ-UHFFFAOYSA-N
diethylaniline,n,n-diethylanilin,diethylphenylamine,benzenamine, n,n-diethyl,n,n-diethyl aniline,diaethylanilin,phenyldiethylamine,aniline, n,n-diethyl,n,n-diethylaminobenzene,n-phenyldiethylamine
7061
N,N-diethylaniline
CCN(CC)C1=CC=CC=C1
91-66-7
C10H15N
149.24
MFCD00009042
GGSUCNLOZRCGPQ-UHFFFAOYSA-N
diethylaniline,n,n-diethylanilin,diethylphenylamine,benzenamine, n,n-diethyl,n,n-diethyl aniline,diaethylanilin,phenyldiethylamine,aniline, n,n-diethyl,n,n-diethylaminobenzene,n-phenyldiethylamine
7061
N,N-diethylaniline
CCN(CC)C1=CC=CC=C1
Specific Gravity 0.933
Refractive Index 1.5410 to 1.5430
Infrared Spectrum Authentic
Quantity 2.5L
Packaging Glass bottle
Flash Point 84°C
Melting Point -38.0°C
Density 0.9330g/mL
Color Yellow
Boiling Point 215.0°C to 217.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H5N(C2H5)2
Beilstein 12, 164
Fieser 05,212; 16,182
Merck Index 15, 3125
Solubility Information Solubility in water: 10-27g/L. Other solubilities: slightly soluble in alcohol,chloroform and ether
Formula Weight 149.24
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material N, N-Diethylaniline, 99%
Show More Show Less

GHS H Statement
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.
Toxic if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre

GHS Signal Word: Danger

EINECSNumber : 202-088-8

RTECSNumber : BX3400000

TSCA : TSCA