Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethylethanolamine, 99%, Thermo Scientific Chemicals

Catalog Number p-109162
Click to view available options
Quantity:
10 L
Packaging:
Plastic drum
100-37-8
MFCD00002850
BFSVOASYOCHEOV-UHFFFAOYSA-N
2-diethylamino ethanol, n,n-diethylethanolamine, diethylaminoethanol, deae, diethylethanolamine, diethylamino ethanol, ethanol, 2-diethylamino, n,n-diethyl-2-aminoethanol, 2-hydroxyethyl diethylamine, diethyl 2-hydroxyethyl amine
7497
CHEBI:52153
CCN(CC)CCO
100-37-8
MFCD00002850
BFSVOASYOCHEOV-UHFFFAOYSA-N
2-diethylamino ethanol, n,n-diethylethanolamine, diethylaminoethanol, deae, diethylethanolamine, diethylamino ethanol, ethanol, 2-diethylamino, n,n-diethyl-2-aminoethanol, 2-hydroxyethyl diethylamine, diethyl 2-hydroxyethyl amine
7497
CHEBI:52153
CCN(CC)CCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -70.0°C
Color White to Yellow
Density 0.8800g/mL
Boiling Point 161.0°C
Flash Point 51.5°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic drum
Molecular Formula C6H15NO
Refractive Index 1.4400 to 1.4420
Linear Formula HOCH2CH2N(C2H5)2
Quantity 10 L
Specific Gravity 0.88
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and benzene
IUPAC Name 2-(diethylamino)ethanol
Molecular Weight (g/mol) 117.192
Formula Weight 117.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N, N-Diethylethanolamine, 99%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement
Harmful in contact with skin.
Toxic if inhaled.
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-845-2
RTECSNumber KK5075000
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit