Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethylformamide, 99%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
500mL
Packaging:
Glass bottle
617-84-5
C5H11NO
101.15
MFCD00003287
SUAKHGWARZSWIH-UHFFFAOYSA-N
diethylformamide,formamide, n,n-diethyl,n-formyldiethylamine,diethyl formamide,formyldiethylamine,diethylamid kyseliny mravenci,n,n-diethylcarboxamide,diethylamid kyseliny mravenci czech,diethyl-formamide,formamide, diethyl
12051
N,N-diethylformamide
CCN(CC)C=O
617-84-5
C5H11NO
101.15
MFCD00003287
SUAKHGWARZSWIH-UHFFFAOYSA-N
diethylformamide,formamide, n,n-diethyl,n-formyldiethylamine,diethyl formamide,formyldiethylamine,diethylamid kyseliny mravenci,n,n-diethylcarboxamide,diethylamid kyseliny mravenci czech,diethyl-formamide,formamide, diethyl
12051
N,N-diethylformamide
CCN(CC)C=O
Flash Point 60°C
Color Colorless to Yellow
Boiling Point 176.0°C to 177.0°C
Density 0.9000g/mL
Quantity 500mL
Infrared Spectrum Authentic
Specific Gravity 0.9
Packaging Glass bottle
Refractive Index 1.4330 to 1.4350
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula HCON(C2H5)2
Beilstein 04, 109
Formula Weight 101.15
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material N, N-Diethylformamide, 99%
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get med

GHS Signal Word: Warning

EINECSNumber : 210-533-2

RTECSNumber : LQ1925000

TSCA : TSCA