Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N,N'-Diethylhydrazine dihydrochloride, tech. 90%

Click to view available options
Quantity:
5g
25g
7699-31-2
C4H14Cl2N2
161.07
MFCD00012523
NMTDWUVFGOBBAX-UHFFFAOYSA-N
1,2-diethylhydrazine dihydrochloride,n,n'-diethylhydrazine dihydrochloride,unii-x3o3iwe4tg,ccris 8919,hydrazine, 1,2-diethyl-, dihydrochloride,x3o3iwe4tg,1,2-diethylhydrazine 2hcl,diethylhydrazine dihydrochloride,1,2-diethyl hydrazine dihydrochloride,1,2-diethylhydrazine dihydrochlo-ride
24372
1,2-diethylhydrazine;dihydrochloride
CCNNCC.Cl.Cl
7699-31-2
C4H14Cl2N2
161.07
MFCD00012523
NMTDWUVFGOBBAX-UHFFFAOYSA-N
1,2-diethylhydrazine dihydrochloride,n,n'-diethylhydrazine dihydrochloride,unii-x3o3iwe4tg,ccris 8919,hydrazine, 1,2-diethyl-, dihydrochloride,x3o3iwe4tg,1,2-diethylhydrazine 2hcl,diethylhydrazine dihydrochloride,1,2-diethyl hydrazine dihydrochloride,1,2-diethylhydrazine dihydrochlo-ride
24372
1,2-diethylhydrazine;dihydrochloride
CCNNCC.Cl.Cl
Quantity 5g
Melting Point ~169°C (decomposition)
Odor Odorless
Beilstein 3668380
Formula Weight 159.06
Percent Purity 90%
Grade Technical
Chemical Name or Material N,N'-Diethylhydrazine dihydrochloride

GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : MV2295000