Alfa Aesar™ N,N'-Diethyloxamide, 98%

Manufacturer: Alfa Aesar™ B2416722

Quantity 100g

 View more versions of this product

Catalog No. AAB2416722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 615-84-9
    Molecular Formula C6H12N2O2
    Formula Weight 144.17
    MDL Number MFCD00043650
    Chemical Name or Material N,N'-Diethyloxamide

    Specifications

    Formula Weight 144.17
    Melting Point 178° to 182°C
    Quantity 100g
    Molecular Formula C6H12N2O2
    Odor Odorless
    MDL Number MFCD00043650
    CAS 615-84-9
    Chemical Name or Material N,N'-Diethyloxamide
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit