Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N,N'-Diethyloxamide, 98%

Click to view available options
Quantity:
25g
100g
615-84-9
C6H12N2O2
144.174
MFCD00043650
FFYAVOJIYAAUNX-UHFFFAOYSA-N
nn'-diethyloxamide,ethanediamide, n,n'-diethyl,n,n'-diethyloxalamide,n,n-diethyloxamide,n,n'-diethylethanediamide,oxamide, n,n'-diethyl,acmc-1bc1a,n,n'-diethyl-oxalamide,n-ethyl-n'-ethylethane-1,2-diamide,n 1 ,n 2-diethylethanediamide
69210
N,N'-diethyloxamide
CCNC(=O)C(=O)NCC
615-84-9
C6H12N2O2
144.174
MFCD00043650
FFYAVOJIYAAUNX-UHFFFAOYSA-N
nn'-diethyloxamide,ethanediamide, n,n'-diethyl,n,n'-diethyloxalamide,n,n-diethyloxamide,n,n'-diethylethanediamide,oxamide, n,n'-diethyl,acmc-1bc1a,n,n'-diethyl-oxalamide,n-ethyl-n'-ethylethane-1,2-diamide,n 1 ,n 2-diethylethanediamide
69210
N,N'-diethyloxamide
CCNC(=O)C(=O)NCC
Quantity 25g
Melting Point 178°C to 182°C
Odor Odorless
Formula Weight 144.17
Percent Purity 98%
Chemical Name or Material N,N'-Diethyloxamide