Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Difluoro-2,2'-bipyridinium bis(tetrafluoroborate), ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
178439-26-4
C10H8B2F10N2
367.792
MFCD01073546
IDFABAACDRPWQX-UHFFFAOYSA-N
1,1'-difluoro-2,2'-bipyridinium bis tetrafluoroborate,1,1'-difluoro-2,2'-bipyridine-1,1'-diium tetrafluoroborate,n,n'-difluoro-2,2'-bipyridinium bis tetrafluoroborate,1-fluoro-2-1-fluoropyridin-1-ium-2-yl pyridin-1-ium ditetrafluoroborate,mec-31,synfluor reagent,1,1'-difluoro-2,2'-bipyridine-1,1'-diium ditetrafluoroborate,daikin mec-31,acmc-209eei,n,n'-difluoro-2,2'-bipyridinium
2733931
1-fluoro-2-(1-fluoropyridin-1-ium-2-yl)pyridin-1-ium;ditetrafluoroborate
[B-](F)(F)(F)F.[B-](F)(F)(F)F.C1=CC=[N+](C(=C1)C2=CC=CC=[N+]2F)F
178439-26-4
C10H8B2F10N2
367.792
MFCD01073546
IDFABAACDRPWQX-UHFFFAOYSA-N
1,1'-difluoro-2,2'-bipyridinium bis tetrafluoroborate,1,1'-difluoro-2,2'-bipyridine-1,1'-diium tetrafluoroborate,n,n'-difluoro-2,2'-bipyridinium bis tetrafluoroborate,1-fluoro-2-1-fluoropyridin-1-ium-2-yl pyridin-1-ium ditetrafluoroborate,mec-31,synfluor reagent,1,1'-difluoro-2,2'-bipyridine-1,1'-diium ditetrafluoroborate,daikin mec-31,acmc-209eei,n,n'-difluoro-2,2'-bipyridinium
2733931
1-fluoro-2-(1-fluoropyridin-1-ium-2-yl)pyridin-1-ium;ditetrafluoroborate
[B-](F)(F)(F)F.[B-](F)(F)(F)F.C1=CC=[N+](C(=C1)C2=CC=CC=[N+]2F)F
C-X bond formation reagent
Quantity 1g
Color Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Formula Weight 367.79
Physical Form Crystalline Powder
Percent Purity 10.0 to 11.5% (effective fluorine) typically
Chemical Name or Material N, N'-Difluoro-2, 2'-bipyridinium bis(tetrafluoroborate)

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger