Alfa Aesar™ N,N-Diisopropyl-1,5-pentanediamine, 97%

Manufacturer: Alfa Aesar™ H5029203

Quantity 1g

 View more versions of this product

Catalog No. AAH5029203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 209803-40-7
    Molecular Formula C11H27N2
    Linear Formula [(CH3)2CH]2NH(CH2)5NH2
    Formula Weight 187.35
    MDL Number MFCD00048285
    UN Number UN2735
    Synonym 5-(Diisopropylamino)amylamine; 5-(Diisopropylamino)pentylamine
    Chemical Name or Material N,N-Diisopropyl-1,5-pentanediamine

    Specifications

    Formula Weight 187.35
    Quantity 1g
    Molecular Formula C11H27N2
    Linear Formula [(CH3)2CH]2NH(CH2)5NH2
    MDL Number MFCD00048285
    UN Number UN2735
    Synonym 5-(Diisopropylamino)amylamine; 5-(Diisopropylamino)pentylamine
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    CAS 209803-40-7
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material N,N-Diisopropyl-1,5-pentanediamine
    Assay 97%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit