Alfa Aesar™ N,N-Diisopropylbenzamide, 99%

Manufacturer: Alfa Aesar™ B2291814

Quantity 25g

 View more versions of this product

Catalog No. AAB2291814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 20383-28-2
    Molecular Formula C13H19NO
    Formula Weight 205.3
    MDL Number MFCD00026370
    Beilstein 2048090
    Chemical Name or Material N,N-Diisopropylbenzamide

    Specifications

    Formula Weight 205.3
    Melting Point 68° to 72°C
    Boiling Point 148° to 152°C (18mmHg)
    Quantity 25g
    Molecular Formula C13H19NO
    MDL Number MFCD00026370
    Beilstein 2048090
    CAS 20383-28-2
    Chemical Name or Material N,N-Diisopropylbenzamide
    Assay 99%

    Description

    Applications
    N,N-Diisopropylbenzamide is used as a pharmaceutical intermediate.

    Solubility
    Soluble in water 301 mg/L @ 25°C.

    Notes
    Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit