Learn More
Chem-Impex International, Inc.Supplier Diversity Partner N,N'-Diisopropylcarbodiimide, ≥ 99% (Assay), CAS#: 693-13-0, 1KG DFS Item
Manufacturer:  Chem-Impex International, Inc.Supplier Diversity Partner 001101KG
N,N'-Diisopropylcarbodiimide, ≥ 99% (Assay), CAS#: 693-13-0, 1KG. Hazardous Material: YES. MDL #: MFCD00065689. Molecular Formula: C7H14N2. Molecular Weight: 126.2. Boiling Point: 145-148 °C. Appearance: Colorless to yellow liquid. Warnings: Highly toxic! Flammable liquid! Storage Temperature: Store at RT. Refractive Index: 1.42-1.44. Density: 0.815 g/mL at 20 °C. Flash Point: 33 ºC. Additional Comments: Used in custom synthesis: peptide coupling, amino acid, protein synthesis. Used in API: dehydrate for amikacin/Fosinopril/Valacyclovir/glutathione dehydrants. Use in oligonucleotides,RNA synthesis. Industry use: special coating & adhesive Chem-Impex charges a Hazardous fee of $23.50 to each order for this item.
Catalog No. 50-490-452