Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Diisopropylcarbodiimide 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
693-13-0
C7H14N2
126.2
MFCD00065689
BDNKZNFMNDZQMI-UHFFFAOYSA-N
n,n'-diisopropylcarbodiimide,diisopropylcarbodiimide,1,3-diisopropylcarbodiimide,n,n-diisopropylcarbodiimide,carbodiimide, diisopropyl,dipcdi,dic,2-propanamine, n,n'-methanetetraylbis,diisopropylmethanediimine,n,n'-methanediylidenebis propan-2-amine
12734
CHEBI:53092
N,N'-di(propan-2-yl)methanediimine
CC(C)N=C=NC(C)C
693-13-0
C7H14N2
126.2
MFCD00065689
BDNKZNFMNDZQMI-UHFFFAOYSA-N
n,n'-diisopropylcarbodiimide,diisopropylcarbodiimide,1,3-diisopropylcarbodiimide,n,n-diisopropylcarbodiimide,carbodiimide, diisopropyl,dipcdi,dic,2-propanamine, n,n'-methanetetraylbis,diisopropylmethanediimine,n,n'-methanediylidenebis propan-2-amine
12734
CHEBI:53092
N,N'-di(propan-2-yl)methanediimine
CC(C)N=C=NC(C)C
Coupling reagent
Specific Gravity 0.81
Density 0.8100g/mL
Quantity 500g
Infrared Spectrum Authentic
Boiling Point 145.0°C to 148.0°C
Flash Point 33°C
Color Colorless to Yellow
Refractive Index 1.4320 to 1.4350
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)2CHN=C=NCH(CH3)2
Merck Index 15,3218
Solubility Information Solubility in water: decomposes
Formula Weight 126.2
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material N, N'-Diisopropylcarbodiimide, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 211-743-7

RTECSNumber : FF2175000

TSCA : TSCA