Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Diisopropylcarbodiimide, 99%, AcroSeal™, Thermo Scientific Chemicals

Catalog Number ac446181000
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
693-13-0
C7H14N2
126.2
MFCD00065689
BDNKZNFMNDZQMI-UHFFFAOYSA-N
n,n'-diisopropylcarbodiimide, diisopropylcarbodiimide, 1,3-diisopropylcarbodiimide, n,n-diisopropylcarbodiimide, carbodiimide, diisopropyl, dipcdi, dic, 2-propanamine, n,n'-methanetetraylbis, diisopropylmethanediimine, n,n'-methanediylidenebis propan-2-amine
12734
CHEBI:53092
N,N'-di(propan-2-yl)methanediimine
CC(C)N=C=NC(C)C
693-13-0
C7H14N2
126.2
MFCD00065689
BDNKZNFMNDZQMI-UHFFFAOYSA-N
n,n'-diisopropylcarbodiimide, diisopropylcarbodiimide, 1,3-diisopropylcarbodiimide, n,n-diisopropylcarbodiimide, carbodiimide, diisopropyl, dipcdi, dic, 2-propanamine, n,n'-methanetetraylbis, diisopropylmethanediimine, n,n'-methanediylidenebis propan-2-amine
12734
CHEBI:53092
N,N'-di(propan-2-yl)methanediimine
CC(C)N=C=NC(C)C

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.8100g/mL
Boiling Point 145.0°C to 148.0°C
Flash Point 33°C
Assay Percent Range 98.5% min. (GC)
Packaging AcroSeal™ Glass bottle
Linear Formula (CH3)2CHN=C=NCH(CH3)2
Refractive Index 1.4320 to 1.4350
Quantity 100 mL
Merck Index 15,3218
Specific Gravity 0.81
Solubility Information Solubility in water: decomposes
Formula Weight 126.2
Percent Purity 99%
Chemical Name or Material N, N'-Diisopropylcarbodiimide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes seriou
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rins
EINECSNumber 211-743-7

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit