Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diisopropylethylamine, 99.5+%, ACROS Organics™

Click to view available options
Quantity:
25mL
50mL
100mL
1L
2.5L
Packaging:
Glass Bottle
7087-68-5
C8H19N
129.24
MFCD00008868
JGFZNNIVVJXRND-UHFFFAOYSA-N
n,n-diisopropylethylamine,ethyldiisopropylamine,n-ethyldiisopropylamine,diisopropylethylamine,diea,hunig's base,n-ethyl-n-isopropylpropan-2-amine,dipea,2-propanamine, n-ethyl-n-1-methylethyl,1,1'-dimethyltriethylamine
81531
N-ethyl-N-propan-2-ylpropan-2-amine
CCN(C(C)C)C(C)C
7087-68-5
C8H19N
129.24
MFCD00008868
JGFZNNIVVJXRND-UHFFFAOYSA-N
n,n-diisopropylethylamine,ethyldiisopropylamine,n-ethyldiisopropylamine,diisopropylethylamine,diea,hunig's base,n-ethyl-n-isopropylpropan-2-amine,dipea,2-propanamine, n-ethyl-n-1-methylethyl,1,1'-dimethyltriethylamine
81531
N-ethyl-N-propan-2-ylpropan-2-amine
CCN(C(C)C)C(C)C
Specific Gravity 0.756
Viscosity 0.88 mm2/s (20°C)
Water 0.1% max. (K.F.)
Packaging Glass Bottle
Infrared Spectrum Authentic
Density 0.7560g/mL
Quantity 1L
Refractive Index 1.4123 to 1.4143
Color Colorless to Yellow
Boiling Point 127.0°C
Flash Point 6°C
Melting Point -46.0°C
CAS Min % 99.5
CAS Max % 100.0
Assay Percent Range 99.5% min. (GC)
Linear Formula [(CH3)2CH]2NC2H5
UN Number 2733
Beilstein 04,IV,511
Fieser 01,371; 04,230; 07,148; 10,151; 12,277; 13,121; 16,176
Merck Index 15,3219
Solubility Information Solubility in water: 3.9g/L (20°C)
Formula Weight 129.24
Physical Form Liquid
Percent Purity 99.5+%
Chemical Name or Material N, N-Diisopropylethylamine, 99.5+%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Toxic if inhaled.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 230-392-