Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diisopropylethylamine 99.0+%, TCI America™

Click to view available options
Quantity:
25mL
100mL
500mL
7087-68-5
C8H19N
129.247
MFCD00008868
JGFZNNIVVJXRND-UHFFFAOYSA-N
n,n-diisopropylethylamine,ethyldiisopropylamine,n-ethyldiisopropylamine,diisopropylethylamine,diea,hunig's base,n-ethyl-n-isopropylpropan-2-amine,dipea,2-propanamine, n-ethyl-n-1-methylethyl,1,1'-dimethyltriethylamine
81531
N-ethyl-N-propan-2-ylpropan-2-amine
CCN(C(C)C)C(C)C
7087-68-5
C8H19N
129.247
MFCD00008868
JGFZNNIVVJXRND-UHFFFAOYSA-N
n,n-diisopropylethylamine,ethyldiisopropylamine,n-ethyldiisopropylamine,diisopropylethylamine,diea,hunig's base,n-ethyl-n-isopropylpropan-2-amine,dipea,2-propanamine, n-ethyl-n-1-methylethyl,1,1'-dimethyltriethylamine
81531
N-ethyl-N-propan-2-ylpropan-2-amine
CCN(C(C)C)C(C)C
Boiling Point 127°C
Color Yellow
Melting Point -45°C
Quantity 25mL
UN Number 2733
Formula Weight 129.25
Physical Form Clear Liquid at 20°C
Percent Purity ≥99.0% (GC)
Chemical Name or Material N,N-Diisopropylethylamine

EINECSNumber : (2)-3499

TSCA : Yes