Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diisopropylethylenediamine, 97%, Thermo Scientific Chemicals

Catalog Number p-4813565
Click to view available options
Quantity:
25 mL
100 mL
121-05-1
C8H20N2
144.26
CURJNMSGPBXOGK-UHFFFAOYSA-N
2-aminoethyldiisopropylamine, n,n-diisopropylethylenediamine, 2-diisopropylamino ethylamine, usaf am-2, 1,2-ethanediamine, n,n-bis 1-methylethyl, n,n-diisopropyl-1,2-ethanediamine, n1,n1-diisopropylethane-1,2-diamine, 2-aminoethyl diisopropylamine, n,n-diisopropyl ethylenediamine, 2-diisopropylaminoethylamine
8459
N',N'-di(propan-2-yl)ethane-1,2-diamine
CC(C)N(CCN)C(C)C
121-05-1
C8H20N2
144.26
CURJNMSGPBXOGK-UHFFFAOYSA-N
2-aminoethyldiisopropylamine, n,n-diisopropylethylenediamine, 2-diisopropylamino ethylamine, usaf am-2, 1,2-ethanediamine, n,n-bis 1-methylethyl, n,n-diisopropyl-1,2-ethanediamine, n1,n1-diisopropylethane-1,2-diamine, 2-aminoethyl diisopropylamine, n,n-diisopropyl ethylenediamine, 2-diisopropylaminoethylamine
8459
N',N'-di(propan-2-yl)ethane-1,2-diamine
CC(C)N(CCN)C(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.8300g/mL
Boiling Point 170°C
Flash Point 54°C
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula [(CH3)2CH]2NCH2CH2NH2
Quantity 25 mL
Specific Gravity 0.83
Solubility Information Solubility in water: miscible
Formula Weight 144.26
Percent Purity 97%
Chemical Name or Material N, N-Diisopropylethylenediamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
EINECSNumber 204-447-4

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit