Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethyl-1-naphthylamine, 99%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
100mL
500mL
Packaging:
Glass bottle
86-56-6
C12H13N
171.24
MFCD00003919
AJUXDFHPVZQOGF-UHFFFAOYSA-N
n,n-dimethyl-1-naphthylamine,1-dimethylaminonaphthalene,1-naphthalenamine, n,n-dimethyl,n,n-dimethyl-1-naphthalenamine,n,n-dimethyl-1-napthylamine,dimethyl 1-naphthyl amine,alpha-dimethylaminonaphthalene,1-naphthylamine, n,n-dimethyl,dimethyl-alpha-naphthylamine,n,n-dimethyl-1-naftylamin
6848
N,N-dimethylnaphthalen-1-amine
CN(C)C1=CC=CC2=CC=CC=C21
86-56-6
C12H13N
171.24
MFCD00003919
AJUXDFHPVZQOGF-UHFFFAOYSA-N
n,n-dimethyl-1-naphthylamine,1-dimethylaminonaphthalene,1-naphthalenamine, n,n-dimethyl,n,n-dimethyl-1-naphthalenamine,n,n-dimethyl-1-napthylamine,dimethyl 1-naphthyl amine,alpha-dimethylaminonaphthalene,1-naphthylamine, n,n-dimethyl,dimethyl-alpha-naphthylamine,n,n-dimethyl-1-naftylamin
6848
N,N-dimethylnaphthalen-1-amine
CN(C)C1=CC=CC2=CC=CC=C21
Infrared Spectrum Authentic
Refractive Index 1.6210 to 1.623
Boiling Point 139°C to 140°C (13.0mmHg)
Flash Point 113°C
Specific Gravity 1.04
Packaging Glass bottle
Quantity 5mL
Color Yellow to Brown
Density 1.0400g/mL
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C10H7N(CH3)2
Beilstein 12, 1221
Merck Index 15, 3276
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 171.24
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material N, N-Dimethyl-1-naphthylamine, 99%
Show More Show Less

GHS H Statement:
Toxic to aquatic life with long lasting effects.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 201-682-4

RTECSNumber : QM2825000

TSCA : TSCA