Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethyl-1-naphthylamine, 99%, Thermo Scientific Chemicals

Catalog Number AC204170250
Click to view available options
Quantity:
5 mL
25 mL
100 mL
500 mL
Packaging:
Glass bottle
86-56-6
C12H13N
171.24
MFCD00003919
AJUXDFHPVZQOGF-UHFFFAOYSA-N
n,n-dimethyl-1-naphthylamine, 1-dimethylaminonaphthalene, 1-naphthalenamine, n,n-dimethyl, n,n-dimethyl-1-naphthalenamine, n,n-dimethyl-1-napthylamine, dimethyl 1-naphthyl amine, alpha-dimethylaminonaphthalene, 1-naphthylamine, n,n-dimethyl, dimethyl-alpha-naphthylamine, n,n-dimethyl-1-naftylamin
6848
N,N-dimethylnaphthalen-1-amine
CN(C)C1=CC=CC2=CC=CC=C21
86-56-6
C12H13N
171.24
MFCD00003919
AJUXDFHPVZQOGF-UHFFFAOYSA-N
n,n-dimethyl-1-naphthylamine, 1-dimethylaminonaphthalene, 1-naphthalenamine, n,n-dimethyl, n,n-dimethyl-1-naphthalenamine, n,n-dimethyl-1-napthylamine, dimethyl 1-naphthyl amine, alpha-dimethylaminonaphthalene, 1-naphthylamine, n,n-dimethyl, dimethyl-alpha-naphthylamine, n,n-dimethyl-1-naftylamin
6848
N,N-dimethylnaphthalen-1-amine
CN(C)C1=CC=CC2=CC=CC=C21

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Yellow to Brown
Density 1.0400g/mL
Boiling Point 139°C to 140°C (13.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.6210 to 1.623
Linear Formula C10H7N(CH3)2
Quantity 25 mL
Beilstein 12, 1221
Specific Gravity 1.04
Merck Index 15, 3276
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 171.24
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Dimethyl-1-naphthylamine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Toxic to aquatic life with long lasting effects.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-682-4
RTECSNumber QM2825000
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit