Alfa Aesar™ N,N-Dimethyl-2-(1-piperazinyl)nicotinamide hydrochloride, 96%

Manufacturer: Alfa Aesar™ H51679MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH51679MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 902836-08-2
    Molecular Formula C12H18N4O·HCl
    Formula Weight 270.76
    MDL Number MFCD12407084
    Synonym N,N-Dimethyl-2-(1-piperazinyl)pyridine-3-carboxamide hydrochloride
    Chemical Name or Material N,N-Dimethyl-2-(1-piperazinyl)nicotinamide hydrochloride

    Specifications

    Formula Weight 270.76
    Quantity 250mg
    Molecular Formula C12H18N4O·HCl
    MDL Number MFCD12407084
    CAS 902836-08-2
    Synonym N,N-Dimethyl-2-(1-piperazinyl)pyridine-3-carboxamide hydrochloride
    Chemical Name or Material N,N-Dimethyl-2-(1-piperazinyl)nicotinamide hydrochloride
    Assay 96%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit