Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethyl-4-nitrosoaniline 99%, ACROS Organics™

Click to view available options
Quantity:
100g
Packaging:
Glass bottle
138-89-6
C8H10N2O
150.181
MFCD00002063
CMEWLCATCRTSGF-UHFFFAOYSA-N
ndma,accelerine,p-nitroso-n,n-dimethylaniline,n,n-dimethyl-p-nitrosoaniline,4-nitroso-n,n-dimethylaniline,4-nitrosodimethylaniline,p-nitrosodimethylanilide,paranitrosodimethylanilide,4-dimethylamino nitrosobenzene,benzenamine, n,n-dimethyl-4-nitroso
8749
CHEBI:59990
N,N-dimethyl-4-nitrosoaniline
CN(C)C1=CC=C(C=C1)N=O
138-89-6
C8H10N2O
150.181
MFCD00002063
CMEWLCATCRTSGF-UHFFFAOYSA-N
ndma,accelerine,p-nitroso-n,n-dimethylaniline,n,n-dimethyl-p-nitrosoaniline,4-nitroso-n,n-dimethylaniline,4-nitrosodimethylaniline,p-nitrosodimethylanilide,paranitrosodimethylanilide,4-dimethylamino nitrosobenzene,benzenamine, n,n-dimethyl-4-nitroso
8749
CHEBI:59990
N,N-dimethyl-4-nitrosoaniline
CN(C)C1=CC=C(C=C1)N=O
Sulfated Ash 0.1% max.
Infrared Spectrum Authentic
Packaging Glass bottle
Color Green
Melting Point 83°C to 86°C
Quantity 100g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula ONC6H4N(CH3)2
Beilstein 12, 677
Fieser 01,746
Merck Index 15, 6725
Formula Weight 150.18
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material N, N-Dimethyl-4-nitrosoaniline, 99%
Show More Show Less

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic if inhaled.
Toxic in contact with skin.
Self-heating; may catch fire.

GHS P Statement:
Keep cool. Protect from sunlight.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 205-343-1

RTECSNumber : BX7175000

TSCA : TSCA