Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethyl-m-phenylenediamine dihydrochloride, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
3575-32-4
C8H12N2·2HCl
209.12
MFCD00012971
BZJPIQKDEGXVFG-UHFFFAOYSA-N
n1,n1-dimethylbenzene-1,3-diamine dihydrochloride,n,n-dimethyl-m-phenylenediamine dihydrochloride,m-dimethylamino aniline,n,n-dimethyl-1,3-phenylenediamine dihydrochloride,1,3-benzenediamine, n,n-dimethyl-, dihydrochloride,n,n-dimethylbenzene-1,3-diamine dihydrochloride,3-amino-n,n-dimethylaniline,1,3-benzenediamine, n1,n1-dimethyl-, hydrochloride 1:2,pubchem14991,acmc-20a5k9
77124
3-N,3-N-dimethylbenzene-1,3-diamine;dihydrochloride
CN(C)C1=CC=CC(=C1)N.Cl.Cl
3575-32-4
C8H12N2·2HCl
209.12
MFCD00012971
BZJPIQKDEGXVFG-UHFFFAOYSA-N
n1,n1-dimethylbenzene-1,3-diamine dihydrochloride,n,n-dimethyl-m-phenylenediamine dihydrochloride,m-dimethylamino aniline,n,n-dimethyl-1,3-phenylenediamine dihydrochloride,1,3-benzenediamine, n,n-dimethyl-, dihydrochloride,n,n-dimethylbenzene-1,3-diamine dihydrochloride,3-amino-n,n-dimethylaniline,1,3-benzenediamine, n1,n1-dimethyl-, hydrochloride 1:2,pubchem14991,acmc-20a5k9
77124
3-N,3-N-dimethylbenzene-1,3-diamine;dihydrochloride
CN(C)C1=CC=CC(=C1)N.Cl.Cl
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 217.0°C
Color Purple to White
Quantity 5g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula (CH3)2NC6H4NH2·2HCl
Beilstein 13, 40
Solubility Information Solubility in water: soluble
Formula Weight 209.12
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material N, N-Dimethyl-m-phenylenediamine dihydrochloride
Show More Show Less

GHS H Statement:
Toxic in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Toxic if inhaled.
Causes serious eye irritation.
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 222-693-