Learn More
N,N-Dimethyl-m-phenylenediamine dihydrochloride, 99%, ACROS Organics™
Specifications
Color | Purple to White |
Melting Point | 217.0°C |
Packaging | Glass bottle |
Infrared Spectrum | Authentic |
Quantity | 100g |
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Assay Percent Range | 98.5% min. (Argentometry) |
Linear Formula | (CH3)2NC6H4NH2·2HCl |
Beilstein | 13, 40 |
Show More |
Safety and Handling
GHS H Statement:
Toxic in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Toxic if inhaled.
Causes serious eye irritation.
Toxic if swallowed.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 222-693-