Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethyl-m-toluidine, 98%, ACROS Organics™

Click to view available options
Quantity:
25mL
Packaging:
Glass bottle
121-72-2
C9H13N
135.21
MFCD00008305
CWOMTHDOJCARBY-UHFFFAOYSA-N
n,n-dimethyl-m-toluidine,benzenamine, n,n,3-trimethyl,dimethyl-m-toluidine,m-methyl-n,n-dimethylaniline,n,n-dimethyl-m-methylaniline,m,n,n-trimethylaniline,n,n-dimethyl-3-methylaniline,dimetil-m-toluidina,n,n,3-trimethylbenzenamine,benzene, 1-dimethylamino-3-methyl
8488
N,N,3-trimethylaniline
CC1=CC(=CC=C1)N(C)C
121-72-2
C9H13N
135.21
MFCD00008305
CWOMTHDOJCARBY-UHFFFAOYSA-N
n,n-dimethyl-m-toluidine,benzenamine, n,n,3-trimethyl,dimethyl-m-toluidine,m-methyl-n,n-dimethylaniline,n,n-dimethyl-m-methylaniline,m,n,n-trimethylaniline,n,n-dimethyl-3-methylaniline,dimetil-m-toluidina,n,n,3-trimethylbenzenamine,benzene, 1-dimethylamino-3-methyl
8488
N,N,3-trimethylaniline
CC1=CC(=CC=C1)N(C)C
Density 0.94g/mL
Refractive Index 1.5490 to 1.5510 (20°C, 589nm)
Infrared Spectrum Authentic
Color Yellow
Packaging Glass bottle
Quantity 25mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3C6H4N(CH3)2
Beilstein 12, 857
Formula Weight 135.21
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material N, N-Dimethyl-m-toluidine, 98%
Show More Show Less

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.
Toxic if inhaled.
Toxic in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

Recommended Storage : product darkens on exposure to air, light and moisture